Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BARHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020063.1 | NP_064447.1 | 1 | 457 | ||
GenBank mRNA | AY338397.1 | - | 1 | 457 | ||
BC126439.1 | - | 1 | 441 | 8992155 | ||
BC126441.1 | - | 1 | 441 | 8992157 |