Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020768.3 | NP_065819.1 | 3 | 1256 | ||
XM_005268492.2 | XP_005268549.1 | 2 | 1647 | |||
XM_005268493.2 | XP_005268550.1 | 2 | 1043 | |||
XM_006714789.2 | XP_006714852.1 | 2 | 1707 | |||
XM_011537671.2 | XP_011535973.1 | 4 | 1542 | |||
XM_011537672.2 | XP_011535974.1 | 2 | 1647 | |||
XM_011537673.2 | XP_011535975.1 | 2 | 1647 | |||
GenBank mRNA | AB037738.1 | - | 2 | 1718 | ||
AK292477.1 | - | 3 | 1297 | |||
BC111962.1 | - | 1 | 649 | 8327430 | ||
BC113435.1 | - | 1 | 649 | 8322487 | ||
CR749266.1 | - | 3 | 1257 |