Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017403.1 | NP_001017403.1 | 2 | 224 | ||
NM_001017404.1 | NP_001017404.1 | 2 | 95 | |||
NM_021636.2 | NP_067649.2 | 2 | 111 | |||
XM_005245404.3 | XP_005245461.1 | 2 | 168 | |||
XM_011509838.2 | XP_011508140.1 | 2 | 169 | |||
XM_011509839.2 | XP_011508141.1 | 2 | 170 | |||
XM_011509840.2 | XP_011508142.1 | 2 | 169 | |||
XM_017001996.1 | XP_016857485.1 | 2 | 170 | |||
GenBank mRNA | AB049405.1 | - | 2 | 109 | ||
AF190501.1 | - | 2 | 95 | |||
AY358119.1 | - | 2 | 152 | |||
BC047905.1 | - | 2 | 94 | 5212508 |