Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022454.3 | NP_071899.1 | 2 | 1441 | ||
GenBank mRNA | AB073988.1 | - | 2 | 1240 | ||
AK025905.1 | - | 2 | 1441 | |||
BC111770.1 | - | 2 | 629 |