Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITIH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001851.2 | NP_001001851.1 | 6, 7 | 936 | ||
NM_030569.6 | NP_085046.5 | 6, 7 | 936 | |||
NM_032817.5 | NP_116206.4 | 2, 3 | 271 | |||
XM_011519713.2 | XP_011518015.1 | 6, 7 | 936 | |||
XM_011519714.2 | XP_011518016.1 | 6, 7 | 936 | |||
GenBank mRNA | AB075833.1 | - | 3, 4 | 477 | ||
AK027375.1 | - | 6, 7 | 873 | |||
AK027831.1 | - | 3, 4 | 455 | |||
AK027849.1 | - | 1, 2 | 120 | |||
AK075381.1 | - | 6, 7 | 909 | |||
AL833203.1 | - | 6, 7 | 781 | |||
AY238437.1 | - | 6, 7 | 832 | |||
AY358426.1 | - | 6, 7 | 901 | |||
BC136439.1 | - | 6, 7 | 909 | |||
CR627109.1 | - | 1, 2 | 5370 | |||
DA833541.1 | - | 1, 2 | 120 | |||
DA839345.1 | - | 1, 2 | 147 | |||
DB079183.1 | - | 2, 3 | 271 |