Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282415.1 | NP_001269344.1 | 5 | 2194 | ||
NM_001282416.1 | NP_001269345.1 | 5 | 2389 | |||
NM_032805.2 | NP_116194.2 | 6 | 2657 | |||
XM_017023791.1 | XP_016879280.1 | 5 | 2693 | |||
XM_017023792.1 | XP_016879281.1 | 6 | 2597 | |||
GenBank mRNA | AK027455.1 | - | 5 | 2385 | ||
AK074736.1 | - | 5 | 2169 | |||
CN366927.1 | - | 1 | 382 |