Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF709 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152601.3 | NP_689814.1 | 2 | 267 | ||
GenBank mRNA | AK290911.1 | - | 2 | 206 | ||
BC136746.1 | - | 2 | 183 | 9020736 | ||
BC136751.1 | - | 2 | 183 | 9020741 | ||
DA493682.1 | - | 2 | 248 |