Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286680.1 | NP_001273609.1 | 5 | 825 | ||
NM_001286681.1 | NP_001273610.1 | 4 | 411 | |||
NM_182795.1 | NP_877724.1 | 4 | 411 | |||
XM_011544360.2 | XP_011542662.1 | 3 | 1047 | |||
XM_011544362.2 | XP_011542664.1 | 5 | 444 | |||
XM_011544363.2 | XP_011542665.1 | 4 | 325 | |||
XM_011544364.2 | XP_011542666.1 | 4 | 734 | |||
XM_017012948.1 | XP_016868437.1 | 5 | 1044 | |||
XM_017012949.1 | XP_016868438.1 | 4 | 1210 | |||
GenBank mRNA | AK094267.1 | - | 4 | 1186 | ||
AY262113.1 | - | 5 | 501 | |||
AY262114.1 | - | 4 | 411 | |||
BC068078.1 | - | 4 | 403 | 6087597 |