Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021030.2 | NP_066358.2 | 4 | 1532 | ||
GenBank mRNA | AB021644.1 | - | 4 | 1482 | ||
AK074832.1 | - | 4 | 1517 | |||
AV726241.1 | - | 1 | 365 | |||
BC136323.1 | - | 4 | 1419 | 9020310 | ||
CD358782.1 | - | 1 | 653 |