Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK122716.1 | - | 12 | 2502 | ||
AK127756.1 | - | 12 | 2338 | |||
BC128588.1 | - | 11 | 2275 | 40128488 | ||
BC143380.1 | - | 13 | 2391 |