Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD11B1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267868.1 | NP_001254797.1 | 9 | 1782 | ||
NM_001267869.1 | NP_001254798.1 | 6 | 1469 | |||
NM_001267870.1 | NP_001254799.1 | 5 | 1309 | |||
NM_001267871.1 | NP_001254800.1 | 6 | 1238 | |||
NM_198533.2 | NP_940935.1 | 8 | 1481 | |||
NM_198704.2 | NP_941993.1 | 6 | 1306 | |||
NM_198705.2 | NP_941994.1 | 6 | 1401 | |||
NM_198706.2 | NP_941995.1 | 8 | 1644 | |||
NM_198707.2 | NP_941996.1 | 7 | 1581 | |||
NM_198708.2 | NP_941997.1 | 7 | 1532 | |||
GenBank mRNA | AK308206.1 | - | 9 | 1523 | ||
AY268351.1 | - | 6 | 1142 | |||
AY268352.1 | - | 8 | 1345 | |||
AY268353.1 | - | 7 | 1305 | |||
AY268354.1 | - | 7 | 1233 | |||
AY313894.1 | - | 8 | 1210 | |||
AY313895.1 | - | 6 | 1047 | |||
AY313896.1 | - | 6 | 979 | |||
BC053546.1 | - | 7 | 1162 | |||
BC072434.1 | - | 6 | 1218 | |||
BC094798.1 | - | 6 | 1225 |