Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207370.2 | NP_997253.2 | 6 | 2360 | ||
XM_011541434.2 | XP_011539736.1 | 6 | 2464 | |||
XM_017001250.1 | XP_016856739.1 | 5 | 2446 | |||
GenBank mRNA | BC068275.1 | - | 6 | 2360 | 5167285 | |
CR627473.1 | - | 1 | 996 |