Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020877.3 | NP_065928.2 | 18 | 4378 | ||
XM_011523663.1 | XP_011521965.1 | 20 | 3246 | |||
XM_011523664.2 | XP_011521966.1 | 21 | 3395 | |||
XM_011523666.1 | XP_011521968.1 | 20 | 3246 | |||
XM_011523667.2 | XP_011521969.1 | 19 | 3149 | |||
XM_011523668.1 | XP_011521970.1 | 10 | 1629 | |||
XM_011523669.2 | XP_011521971.1 | 8 | 1313 | |||
XM_011523670.2 | XP_011521972.1 | 20 | 3246 | |||
XM_017024218.1 | XP_016879707.1 | 7 | 1330 | |||
XM_017024219.1 | XP_016879708.1 | 20 | 3246 | |||
GenBank mRNA | AB040936.2 | - | 18 | 4378 | ||
BX648163.1 | - | 1 | 132 |