Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099456.2 | NP_001092926.2 | 1 | 701 | ||
GenBank mRNA | AB084276.1 | - | 1 | 481 | ||
AL365519.1 | - | 1 | 21 | |||
BC150534.1 | - | 1 | 205 | |||
BC171886.1 | - | 1 | 918 |