Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB40C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172663.1 | NP_001166134.1 | 7 | 1533 | ||
NM_001172664.1 | NP_001166135.1 | 7 | 1425 | |||
NM_001172665.1 | NP_001166136.1 | 7 | 1385 | |||
NM_001172666.1 | NP_001166137.1 | 7 | 1328 | |||
NM_021168.4 | NP_066991.3 | 6 | 1430 | |||
GenBank mRNA | BC028696.1 | - | 6 | 1432 | 4826800 |