Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300985.1 | NP_001287914.1 | 3 | 332 | ||
NM_001300986.1 | NP_001287915.1 | 3 | 356 | |||
NM_001300987.1 | NP_001287916.1 | 4 | 488 | |||
NM_001300988.1 | NP_001287917.1 | 3 | 332 | |||
NM_001300989.1 | NP_001287918.1 | 3 | 332 | |||
NM_021190.3 | NP_067013.1 | 3 | 332 | |||
NR_125356.1 | 3 | 224 | ||||
NR_125357.1 | 3 | 332 | ||||
XM_011541874.2 | XP_011540176.1 | 3 | 165 | |||
XM_011541875.2 | XP_011540177.1 | 3 | 178 | |||
XM_011541876.2 | XP_011540178.1 | 3 | 195 | |||
XM_017001951.1 | XP_016857440.1 | 2 | 195 | |||
GenBank mRNA | AB051232.1 | - | 3 | 154 | ||
AB051233.1 | - | 3 | 98 | |||
AB209266.1 | - | 3 | 138 | |||
AF176085.1 | - | 3 | 124 | |||
AF530580.1 | - | 3 | 127 | |||
AF530581.1 | - | 3 | 127 | |||
AK295384.1 | - | 3 | 145 | |||
AK299755.1 | - | 3 | 153 | |||
AK299897.1 | - | 4 | 269 | |||
AK299922.1 | - | 3 | 170 | |||
BC016582.2 | - | 3 | 101 | 3872881 | ||
BP250235.1 | - | 3 | 224 | |||
DB473562.1 | - | 3 | 322 |