Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVOL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303461.1 | NP_001290390.1 | 3 | 636 | ||
NM_001303462.1 | NP_001290391.1 | 2 | 225 | |||
NM_021220.3 | NP_067043.2 | 3 | 685 | |||
GenBank mRNA | AK022284.1 | - | 3 | 680 | ||
AL079276.1 | - | 2 | 265 | |||
BC006148.1 | - | 3 | 685 | 3959436 | ||
BE791524.1 | - | 3 | 498 | |||
BM563561.1 | - | 2 | 225 | |||
BT007295.1 | - | 3 | 437 |