Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEZ6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184773.1 | NP_001171702.1 | 6 | 1694 | ||
NM_001184774.1 | NP_001171703.1 | 6 | 1694 | |||
NM_001184775.1 | NP_001171704.1 | 6 | 1694 | |||
NM_001184776.1 | NP_001171705.1 | 6 | 1694 | |||
NM_001184777.1 | NP_001171706.1 | 6 | 1694 | |||
NM_021115.4 | NP_066938.2 | 6 | 1694 | |||
XM_005261439.3 | XP_005261496.1 | 6 | 1670 | |||
XM_005261440.3 | XP_005261497.1 | 6 | 1671 | |||
XM_006724195.3 | XP_006724258.1 | 6 | 1670 | |||
XM_011530037.2 | XP_011528339.1 | 6 | 1670 | |||
XM_011530038.2 | XP_011528340.1 | 6 | 1670 | |||
XM_011530039.2 | XP_011528341.1 | 6 | 1670 | |||
XM_017028699.1 | XP_016884188.1 | 6 | 1671 | |||
XM_017028700.1 | XP_016884189.1 | 6 | 1671 | |||
GenBank mRNA | AB023144.2 | - | 6 | 1654 | ||
AB041736.1 | - | 6 | 1498 | |||
AL035545.1 | - | 5 | 1559 | |||
AL050253.1 | - | 5 | 1559 | |||
AY358405.1 | - | 6 | 1694 | |||
BC126115.1 | - | 6 | 1593 | 8991831 | ||
BC143851.1 | - | 6 | 1593 | 9052367 | ||
BC143854.1 | - | 6 | 1593 | 9052370 | ||
CR456574.1 | - | 6 | 1510 |