Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021174.5 | NP_066997.3 | 8 | 1092 | ||
NR_033902.1 | 8 | 910 | ||||
XM_011544603.1 | XP_011542905.1 | 8 | 910 | |||
XM_017013717.1 | XP_016869206.1 | 8 | 1092 | |||
GenBank mRNA | AB075847.1 | - | 5 | 401 | ||
AK095712.1 | - | 8 | 766 | |||
AK314535.1 | - | 8 | 785 | |||
AL834162.1 | - | 8 | 1079 | |||
AL834351.1 | - | 8 | 1080 | |||
AL834352.1 | - | 8 | 1080 | |||
BC065495.1 | - | 8 | 768 | 5496068 | ||
BX390251.1 | - | 3 | 276 | |||
BX640952.1 | - | 8 | 779 |