Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021101.4 | NP_066924.1 | 4 | 2702 | ||
GenBank mRNA | AF101051.1 | - | 4 | 2681 | ||
AK225963.1 | - | 2 | 1473 | |||
AY358652.1 | - | 4 | 2680 |