Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021174.5 | NP_066997.3 | 13 | 2029 | ||
NR_033902.1 | 13 | 1847 | ||||
XM_011544603.1 | XP_011542905.1 | 13 | 1847 | |||
XM_011544604.1 | XP_011542906.1 | 12 | 1727 | |||
XM_017013717.1 | XP_016869206.1 | 13 | 2029 | |||
GenBank mRNA | AB075847.1 | - | 10 | 1338 | ||
AF293335.1 | - | 2 | 285 | |||
AK095712.1 | - | 13 | 1703 | |||
AK096547.1 | - | 4 | 678 | |||
AK314535.1 | - | 13 | 1722 | |||
AL834162.1 | - | 13 | 2016 | |||
AL834351.1 | - | 13 | 2017 | |||
AL834352.1 | - | 13 | 2017 | |||
BC018269.1 | - | 1 | 51 | |||
BC065495.1 | - | 13 | 1705 | 5496068 | ||
BX640952.1 | - | 13 | 1716 |