Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRHL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195010.1 | NP_001181939.1 | 16 | 2411 | ||
NM_021180.3 | NP_067003.2 | 16 | 2307 | |||
NM_198173.2 | NP_937816.1 | 16 | 2476 | |||
XM_011541869.1 | XP_011540171.1 | 16 | 2357 | |||
XM_011541870.2 | XP_011540172.1 | 15 | 2278 | |||
GenBank mRNA | AL137763.1 | - | 4 | 792 | ||
BC036890.2 | - | 16 | 2422 | 5502632 |