Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021194.2 | NP_067017.2 | 2 | 1254 | ||
GenBank mRNA | AB451467.1 | - | 2 | 1109 | ||
BC121015.1 | - | 2 | 1239 | 40117054 | ||
BC121016.1 | - | 2 | 1239 | 40117055 | ||
CB990012.1 | - | 1 | 466 |