Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SQRDL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271213.1 | NP_001258142.1 | 7 | 873 | ||
NM_021199.3 | NP_067022.1 | 6 | 1066 | |||
GenBank mRNA | AF042284.1 | - | 6 | 756 | ||
AF118085.1 | - | 5 | 587 | |||
AF151802.1 | - | 6 | 750 | |||
AK130140.1 | - | 7 | 873 | |||
AK222463.1 | - | 6 | 765 | |||
AK313111.1 | - | 6 | 745 | |||
BC016836.1 | - | 6 | 721 | 3884024 |