Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM35A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021637.2 | NP_067650.1 | 1 | 266 | ||
GenBank mRNA | AK024146.1 | - | 1 | 239 | ||
AK223240.1 | - | 1 | 234 | |||
AK297745.1 | - | 1 | 231 | |||
BC050273.1 | - | 1 | 231 | 4801325 | ||
BC078658.1 | - | 1 | 231 | 5261160 |