Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172732.1 | NP_001166203.1 | 3 | 656 | ||
NM_001172739.1 | NP_001166210.1 | 3 | 1000 | |||
NM_001172743.1 | NP_001166214.1 | 3 | 1003 | |||
NM_021785.4 | NP_068557.3 | 2 | 806 | |||
NR_033348.1 | 3 | 1079 | ||||
NR_033349.1 | 2 | 885 | ||||
XM_006724459.2 | XP_006724522.1 | 4 | 1155 | |||
XM_006724460.1 | XP_006724523.1 | 3 | 903 | |||
XM_011545439.2 | XP_011543741.1 | 4 | 1184 | |||
XM_011545440.2 | XP_011543742.1 | 4 | 1187 | |||
XM_011545441.2 | XP_011543743.1 | 3 | 990 | |||
GenBank mRNA | AK056214.1 | - | 3 | 899 | ||
AK298873.1 | - | 3 | 617 | |||
AK314631.1 | - | 2 | 769 | |||
BC027937.1 | - | 3 | 959 | 5201821 |