Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316887.1 | NP_001303816.1 | 4 | 376 | ||
NM_001316888.1 | NP_001303817.1 | 3 | 404 | |||
NM_001316889.1 | NP_001303818.1 | 2 | 286 | |||
NM_021796.3 | NP_068568.1 | 3 | 358 | |||
XM_011531257.2 | XP_011529559.1 | 3 | 284 | |||
XM_011531260.2 | XP_011529562.1 | 3 | 398 | |||
XM_017029236.1 | XP_016884725.1 | 2 | 326 | |||
GenBank mRNA | AF234654.1 | - | 3 | 363 | ||
AK075086.1 | - | 3 | 358 | |||
BC022335.1 | - | 3 | 343 | 4769552 | ||
BC066327.1 | - | 2 | 286 | 30339131 |