Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282719.1 | NP_001269648.1 | 2 | 176 | ||
NM_001282720.1 | NP_001269649.1 | 2 | 176 | |||
NM_001282723.1 | NP_001269652.1 | 2 | 176 | |||
NM_001282724.1 | NP_001269653.1 | 2 | 176 | |||
NM_021925.3 | NP_068744.1 | 2 | 176 | |||
XM_011533014.2 | XP_011531316.1 | 2 | 185 | |||
GenBank mRNA | AK025473.1 | - | 2 | 114 | ||
AK299244.1 | - | 2 | 117 | |||
AK299555.1 | - | 2 | 147 | |||
AK301493.1 | - | 2 | 147 | |||
AK316310.1 | - | 2 | 147 | |||
BC017473.1 | - | 2 | 133 | 3611020 | ||
DC421670.1 | - | 2 | 176 |