Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021257.3 | NP_067080.1 | 2, 3 | 584 | ||
GenBank mRNA | AF422797.1 | - | 2, 3 | 584 | ||
AJ245946.1 | - | 2, 3 | 209 | |||
AK098350.1 | - | 2, 3 | 186 | |||
AK315395.1 | - | 2, 3 | 297 | |||
BC032509.1 | - | 2, 3 | 291 | 5247233 |