Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037126.1 | NP_001032203.1 | 2 | 162 | ||
NM_021807.3 | NP_068579.3 | 2 | 162 | |||
XM_005250523.4 | XP_005250580.1 | 2 | 146 | |||
XM_017012495.1 | XP_016867984.1 | 2 | 146 | |||
GenBank mRNA | AB051486.1 | - | 2 | 104 | ||
AF380839.1 | - | 2 | 159 | |||
AK027688.1 | - | 2 | 156 | |||
AK295404.1 | - | 2 | 170 | |||
AL831989.2 | - | 2 | 157 | |||
BC026174.1 | - | 2 | 138 | |||
BC067263.1 | - | 2 | 139 | 6183004 | ||
BE244117.1 | - | 2 | 185 | |||
BE246205.1 | - | 2 | 185 |