Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126128.1 | NP_001119600.1 | 2 | 329 | ||
NM_021935.3 | NP_068754.1 | 2 | 329 | |||
XM_017006974.1 | XP_016862463.1 | 2 | 263 | |||
GenBank mRNA | AF182069.2 | - | 2 | 160 | ||
AF333025.1 | - | 2 | 184 | |||
AY349131.1 | - | 2 | 175 | |||
BC069395.1 | - | 2 | 184 | 7262718 | ||
BC096695.1 | - | 2 | 184 | 40014094 | ||
BC098110.1 | - | 2 | 184 | 40014092 | ||
BC098162.1 | - | 2 | 184 | 40014096 |