Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRKACB SIRNA Assays ›
Gene Symbol:
PRKACB
Gene Name:
protein kinase cAMP-activated catalytic subunit beta
Gene Aliases:
PKA C-beta, PKACB
Chromosome Location:
Chr.1: 84077975 - 84238498 on Build GRCh38
UniGene ID:
Hs.487325
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
294
Validated
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRKACB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242857.2
NP_001229786.1
6
395
NM_001242858.2
NP_001229787.1
3
338
NM_001242859.2
NP_001229788.1
5
386
NM_001242860.2
NP_001229789.1
6
530
NM_001242861.2
NP_001229790.1
5
521
NM_001242862.2
NP_001229791.1
2
134
NM_001300915.1
NP_001287844.1
6
530
NM_001300916.1
NP_001287845.1
3
400
NM_001300917.1
NP_001287846.1
3
338
NM_002731.3
NP_002722.1
3
496
NM_182948.3
NP_891993.1
3
400
NM_207578.2
NP_997461.1
3
496
XM_005271015.3
XP_005271072.1
5
347
XM_005271016.1
XP_005271073.1
5
472
XM_005271017.3
XP_005271074.1
4
337
XM_005271018.4
XP_005271075.1
4
657
XM_005271019.1
XP_005271076.1
3
413
XM_005271020.1
XP_005271077.1
3
380
XM_006710758.1
XP_006710821.1
5
486
XM_017001706.1
XP_016857195.1
6
390
XM_017001707.1
XP_016857196.1
6
390
XM_017001708.1
XP_016857197.1
6
491
XM_017001709.1
XP_016857198.1
5
188
XM_017001710.1
XP_016857199.1
5
486
XM_017001711.1
XP_016857200.1
5
188
XM_017001712.1
XP_016857201.1
5
486
XM_017001713.1
XP_016857202.1
3
438
XM_017001714.1
XP_016857203.1
5
472
XM_017001715.1
XP_016857204.1
5
472
XM_017001716.1
XP_016857205.1
4
340
XM_017001717.1
XP_016857206.1
4
661
XM_017001718.1
XP_016857207.1
3
315
XM_017001719.1
XP_016857208.1
3
413
XM_017001720.1
XP_016857209.1
3
413
GenBank mRNA
AB209189.1
-
6
466
AB451239.1
-
3
145
AB451364.1
-
3
145
AK091420.1
-
5
195
AK296482.1
-
6
197
AK304322.1
-
3
325
AK304375.1
-
3
140
AK314550.1
-
3
392
BC016285.1
-
3
327
3916100
BC035058.1
-
3
414
5262372
BI598037.1
-
3
210
BM905428.1
-
3
282
BX485407.1
-
3
381
BX537705.1
-
3
377
BX641026.1
-
3
376
BX647515.1
-
6
475
CR936631.1
-
5
189
DA228647.1
-
2
135
DC322547.1
-
6
396
DC418574.1
-
3
400
M34181.1
-
3
192
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001077645, XM_006233452, XM_006233451, XM_006233453, D10770.
Matching Taqman Gene Expression Assay:
Hs00176944_m1
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
stimulatory C-type lectin receptor signaling pathway
renal water homeostasis
protein phosphorylation
signal transduction
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
blood coagulation
activation of protein kinase A activity
lipoprotein metabolic process
negative regulation of meiotic cell cycle
regulation of protein processing
cellular response to glucagon stimulus
response to clozapine
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
magnesium ion binding
protein serine/threonine kinase activity
cAMP-dependent protein kinase activity
protein binding
ATP binding
kinase activity
ubiquitin protein ligase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0