Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126121.1 | NP_001119593.1 | 6, 7 | 863 | ||
NM_001126122.1 | NP_001119594.1 | 5, 6 | 773 | |||
NM_021734.4 | NP_068380.3 | 6, 7 | 847 | |||
XM_005257559.3 | XP_005257616.1 | 6, 7 | 1485 | |||
XM_005257560.2 | XP_005257617.1 | 6, 7 | 1039 | |||
XM_005257561.3 | XP_005257618.1 | 6, 7 | 1375 | |||
XM_005257562.2 | XP_005257619.1 | 6, 7 | 1022 | |||
XM_006722007.2 | XP_006722070.1 | 6, 7 | 895 | |||
XM_017024926.1 | XP_016880415.1 | 5, 6 | 1437 | |||
XM_017024927.1 | XP_016880416.1 | 4, 5 | 1111 | |||
GenBank mRNA | AF182404.1 | - | 4, 5 | 639 | ||
AJ251857.1 | - | 4, 5 | 677 | |||
AK075239.1 | - | 7, 8 | 869 | |||
AK097882.1 | - | 4, 5 | 1549 | |||
AK225881.1 | - | 4, 5 | 676 | |||
AL833387.1 | - | 6, 7 | 814 | |||
AY346372.1 | - | 3, 4 | 468 | |||
BC001075.2 | - | 6, 7 | 775 | 2821142 | ||
BC005120.1 | - | 6, 7 | 775 | 2821142 | ||
BM455504.1 | - | 3, 4 | 381 |