Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC90B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286116.1 | NP_001273045.1 | 7, 8 | 810 | ||
NM_001286117.1 | NP_001273046.1 | 7, 8 | 1357 | |||
NM_001286118.1 | NP_001273047.1 | 6, 7 | 933 | |||
NM_001286119.1 | NP_001273048.1 | 7, 8 | 1018 | |||
NM_001286120.1 | NP_001273049.1 | 5, 6 | 813 | |||
NM_021825.4 | NP_068597.2 | 7, 8 | 1037 | |||
XM_005274154.2 | XP_005274211.1 | 6, 7 | 1192 | |||
XM_005274156.3 | XP_005274213.1 | 7, 8 | 1024 | |||
XM_005274159.3 | XP_005274216.1 | 8, 9 | 703 | |||
XM_017018103.1 | XP_016873592.1 | 6, 7 | 635 | |||
XM_017018104.1 | XP_016873593.1 | 8, 9 | 1128 | |||
GenBank mRNA | AF182412.1 | - | 7, 8 | 607 | ||
AF182424.1 | - | 7, 8 | 607 | |||
AF271782.1 | - | 7, 8 | 964 | |||
AK055972.1 | - | 7, 8 | 1354 | |||
AK225726.1 | - | 7, 8 | 618 | |||
AK304769.1 | - | 7, 8 | 770 | |||
AK315787.1 | - | 7, 8 | 998 | |||
AL136791.1 | - | 7, 8 | 757 | |||
AV756714.1 | - | 6, 7 | 573 | |||
BC014573.1 | - | 7, 8 | 647 | 3680689 | ||
BC020783.1 | - | 7, 8 | 608 | 4773466 | ||
BC067764.1 | - | 7, 8 | 928 | |||
BC107755.1 | - | 7, 8 | 778 | 6737596 |