Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASTKD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021826.4 | NP_068598.1 | 2 | 749 | ||
GenBank mRNA | AB058695.1 | - | 1 | 3115 | ||
AK023211.1 | - | 2 | 728 | |||
AK301748.1 | - | 2 | 701 | |||
BC007413.2 | - | 2 | 749 | 3010170 | ||
BC011856.1 | - | 2 | 713 | 4298806 |