Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKX3-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256339.1 | NP_001243268.1 | 3 | 499 | ||
NM_006167.3 | NP_006158.2 | 2 | 772 | |||
GenBank mRNA | AF247704.1 | - | 2 | 772 | ||
AF249670.1 | - | 2 | 587 | |||
AF249671.1 | - | 3 | 527 | |||
AF249672.1 | - | 3 | 509 | |||
U80669.1 | - | 2 | 753 |