Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCL1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098725.1 | NP_001092195.1 | 4 | 612 | ||
NM_021966.2 | NP_068801.1 | 4 | 617 | |||
NR_049726.1 | 5 | 841 | ||||
GenBank mRNA | BC003574.1 | - | 4 | 524 | 3544215 | |
BC005831.2 | - | 4 | 524 | 2966563 | ||
BC014024.1 | - | 4 | 528 | 4308655 | ||
BG718395.1 | - | 4 | 617 | |||
X82240.1 | - | 4 | 532 |