Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM204A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134672.1 | NP_001128144.1 | 5, 6 | 511 | ||
NM_022063.2 | NP_071346.1 | 6, 7 | 712 | |||
XM_005270024.1 | XP_005270081.1 | 7, 8 | 889 | |||
GenBank mRNA | AK023250.1 | - | 6, 7 | 691 | ||
AK026630.1 | - | 6, 7 | 705 | |||
BC023577.2 | - | 6, 7 | 695 | 4765095 | ||
BM837313.1 | - | 5, 6 | 504 | |||
CR457304.1 | - | 4, 5 | 445 |