Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIPAS39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193314.1 | NP_001180243.1 | 20 | 2856 | ||
NM_001193315.1 | NP_001180244.1 | 20 | 2456 | |||
NM_001193316.1 | NP_001180245.1 | 19 | 2309 | |||
NM_001193317.1 | NP_001180246.1 | 20 | 2633 | |||
NM_022067.3 | NP_071350.2 | 21 | 2727 | |||
XM_011537066.1 | XP_011535368.1 | 19 | 2354 | |||
XM_017021579.1 | XP_016877068.1 | 19 | 2488 | |||
GenBank mRNA | AK022769.1 | - | 20 | 2404 | ||
AK022925.1 | - | 20 | 2382 | |||
AK024661.1 | - | 9 | 1552 | |||
AK098254.1 | - | 20 | 2817 | |||
AK222706.1 | - | 20 | 2402 | |||
BC015054.1 | - | 21 | 2648 | 3920459 |