Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPNPEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022098.3 | NP_071381.1 | 8 | 1327 | ||
GenBank mRNA | AK313770.1 | - | 8 | 1288 | ||
AL365514.1 | - | 8 | 1310 | |||
AL834310.1 | - | 9 | 1369 | |||
BC001681.2 | - | 8 | 1294 | 2823656 | ||
BC004989.1 | - | 8 | 1294 | 2823656 | ||
BX648018.1 | - | 8 | 1322 | |||
CR456442.1 | - | 8 | 1263 | |||
CR457396.1 | - | 8 | 1226 |