Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CENPK SIRNA Assays ›
Gene Symbol:
CENPK
Gene Name:
centromere protein K
Gene Aliases:
AF5alpha, CENP-K, FKSG14, P33, Solt
Chromosome Location:
Chr.5: 65488919 - 65563194 on Build GRCh38
UniGene ID:
Hs.529778
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
29861
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CENPK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001267038.1
NP_001253967.1
7
519
NM_022145.4
NP_071428.2
7
553
XM_011543534.2
XP_011541836.1
7
497
XM_011543535.2
XP_011541837.1
7
615
XM_011543536.2
XP_011541838.1
8
775
XM_011543537.2
XP_011541839.1
7
531
XM_011543538.2
XP_011541840.1
7
497
XM_011543539.2
XP_011541841.1
7
579
XM_011543540.2
XP_011541842.1
7
497
XM_011543541.2
XP_011541843.1
4
299
XM_011543542.2
XP_011541844.1
4
419
XM_017009690.1
XP_016865179.1
7
758
XM_017009691.1
XP_016865180.1
7
509
XM_017009692.1
XP_016865181.1
7
727
XM_017009693.1
XP_016865182.1
7
497
XM_017009694.1
XP_016865183.1
8
726
XM_017009695.1
XP_016865184.1
6
395
XM_017009696.1
XP_016865185.1
7
497
XM_017009697.1
XP_016865186.1
6
429
XM_017009698.1
XP_016865187.1
6
448
XM_017009699.1
XP_016865188.1
7
531
XM_017009700.1
XP_016865189.1
6
2863
XM_017009701.1
XP_016865190.1
5
494
GenBank mRNA
AF315941.1
-
7
509
AY009151.1
-
5
383
BC005400.1
-
7
509
3839409
BC008504.1
-
7
553
3511065
BG502832.1
-
7
555
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
sister chromatid cohesion
CENP-A containing nucleosome assembly
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0