Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302378.1 | NP_001289307.1 | 3, 4 | 611 | ||
NM_022340.3 | NP_071735.2 | 4, 5 | 767 | |||
XM_005265384.4 | XP_005265441.1 | 4, 5 | 730 | |||
XM_005265385.4 | XP_005265442.1 | 3, 4 | 585 | |||
XM_017007023.1 | XP_016862512.1 | 3, 4 | 603 | |||
XM_017007026.1 | XP_016862515.1 | 3, 4 | 617 | |||
XM_017007027.1 | XP_016862516.1 | 4, 5 | 758 | |||
XM_017007028.1 | XP_016862517.1 | 4, 5 | 758 | |||
GenBank mRNA | AB209673.1 | - | 4, 5 | 727 | ||
AK291288.1 | - | 3, 4 | 602 | |||
AK301735.1 | - | 3, 4 | 603 | |||
AL833545.1 | - | 4, 5 | 759 | |||
AY009133.1 | - | 1, 2 | 144 | |||
BC106940.2 | - | 1, 2 | 144 | 40034008 |