Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022344.3 | NP_071739.2 | 3 | 394 | ||
XM_005258022.3 | XP_005258079.1 | 4 | 412 | |||
XM_017024940.1 | XP_016880429.1 | 3 | 263 | |||
XM_017024941.1 | XP_016880430.1 | 3 | 261 | |||
GenBank mRNA | AF052093.1 | - | 3 | 321 | ||
AF305686.1 | - | 3 | 350 | |||
BC026017.2 | - | 3 | 340 | 3352761 | ||
BC035715.1 | - | 3 | 348 | 5588164 | ||
BC047325.1 | - | 3 | 334 | 5493477 | ||
BC054035.1 | - | 3 | 328 | 6143106 | ||
BP308900.1 | - | 3 | 379 | |||
CD706845.1 | - | 3 | 398 |