Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543544.1 | XP_011541846.1 | 2 | 566 | ||
GenBank mRNA | AB109031.1 | - | 2 | 533 | ||
AB163917.1 | - | 2 | 533 | |||
AF191545.1 | - | 1 | 505 | |||
AK074281.1 | - | 2 | 610 | |||
AK312864.1 | - | 2 | 546 | |||
AY028805.1 | - | 1 | 463 | |||
BC065240.1 | - | 2 | 527 | 6166289 |