Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EEF1A1 SIRNA Assays ›
Gene Symbol:
EEF1A1
Gene Name:
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases:
CCS-3, CCS3, EE1A1, EEF-1, EEF1A, EF-Tu, EF1A, GRAF-1EF, HNGC:16303, LENG7, PTI1, eEF1A-1
Chromosome Location:
Chr.6: 73515750 - 73521032 on Build GRCh38
UniGene ID:
Hs.535192
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
2991
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EEF1A1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001402.5
NP_001393.1
5
824
XM_011535514.2
XP_011533816.1
5
1459
GenBank mRNA
AF174496.1
-
4
654
AF328730.1
-
1
532
AF397403.1
-
3
569
AJ420488.1
-
4
783
AK024888.1
-
1
822
AK026650.1
-
1
824
AK055058.1
-
5
824
AK058051.1
-
5
824
AK058163.1
-
4
642
AK098510.1
-
5
824
AK098518.1
-
5
684
AK098623.1
-
5
824
AK127384.1
-
5
824
AK222515.1
-
1
824
AK222519.1
-
1
473
AK222523.1
-
1
824
AK222551.1
-
1
824
AK222982.1
-
1
824
AK223030.1
-
1
821
AK223046.1
-
1
827
AK289374.1
-
5
824
AK289442.1
-
5
762
AK290691.1
-
5
824
AK297878.1
-
6
622
AK304214.1
-
6
784
AK315827.1
-
5
815
AY043301.1
-
4
1171
BC008587.1
-
1
810
3447654
BC009733.1
-
1
774
3877516
BC009875.2
-
1
814
3948601
BC010735.1
-
1
803
3898364
BC012891.1
-
1
802
3858483
BC014224.2
-
1
813
4335756
BC014377.1
-
1
236
BC014892.1
-
1
120
BC018150.1
-
1
810
3851939
BC018641.2
-
1
810
3660140
BC018850.2
-
1
813
BC019050.2
-
1
776
BC020477.2
-
1
473
BC021686.1
-
1
829
3930863
BC022412.1
-
1
125
BC028674.1
-
1
828
4107346
BC029337.1
-
1
798
BC029343.1
-
1
796
BC029997.1
-
1
811
BC035877.1
-
1
825
BC038339.1
-
1
810
3461588
BC057391.1
-
1
779
4339606
BC063511.1
-
1
247
BC065761.1
-
1
129
BC066893.1
-
1
824
4818863
BC071619.1
-
2
740
4448926
BC071727.1
-
1
779
4514479
BC071741.1
-
1
804
3885539
BC071841.1
-
1
799
4587981
BC072385.1
-
1
811
5585092
BC082268.1
-
1
803
6729595
BC094687.1
-
2
716
30366186
BC111051.1
-
1
814
5585079
DQ867534.1
-
1
510
EF219140.1
-
5
984
EF362804.1
-
5
1054
M29548.1
-
2
356
X03558.1
-
5
814
X16869.1
-
4
776
Back To Top
More Information
Panther Classification:
Molecular Function -
translation factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
translational elongation
cellular response to epidermal growth factor stimulus
regulation of chaperone-mediated autophagy
tRNA binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
protein kinase binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0