Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205319.1 | NP_001192248.1 | 7, 8 | 870 | ||
NM_022463.4 | NP_071908.2 | 7, 8 | 1213 | |||
XM_005256756.4 | XP_005256813.1 | 6, 7 | 1039 | |||
XM_005256758.3 | XP_005256815.1 | 7, 8 | 1542 | |||
GenBank mRNA | AF086523.1 | - | 1, 2 | 41 | ||
AK022676.1 | - | 5, 6 | 642 | |||
AK027451.1 | - | 6, 7 | 766 | |||
AK297990.1 | - | 3, 4 | 342 | |||
AK302073.1 | - | 7, 8 | 872 | |||
AY189690.1 | - | 6, 7 | 753 | |||
BC009327.2 | - | 7, 8 | 807 | |||
BC063828.1 | - | 4, 5 | 468 | |||
BC073845.1 | - | 5, 6 | 623 |