Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022475.2 | NP_071920.1 | 1 | 792 | ||
XM_005263178.4 | XP_005263235.1 | 1 | 661 | |||
XM_006714288.3 | XP_006714351.1 | 1 | 661 | |||
GenBank mRNA | AK074711.1 | - | 1 | 408 | ||
AK291849.1 | - | 1 | 132 | |||
AY009317.1 | - | 1 | 632 | |||
AY009951.1 | - | 1 | 767 | |||
AY358747.1 | - | 1 | 742 | |||
BC009298.2 | - | 1 | 589 | |||
BC025311.1 | - | 1 | 618 | 4122618 |