Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300913.1 | NP_001287842.1 | 4 | 569 | ||
NM_022338.3 | NP_071733.1 | 4 | 569 | |||
XM_005274053.3 | XP_005274110.1 | 4 | 744 | |||
XM_011545103.2 | XP_011543405.1 | 2 | 164 | |||
GenBank mRNA | AF264781.1 | - | 4 | 529 | ||
AF370372.1 | - | 3 | 437 | |||
AY358754.1 | - | 2 | 131 | |||
BC011765.2 | - | 3 | 418 | 3614861 | ||
BG701401.1 | - | 4 | 574 | |||
BI862360.1 | - | 4 | 543 | |||
BX384793.2 | - | 3 | 371 |