Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022346.4 | NP_071741.2 | 2 | 543 | ||
NR_073124.1 | 2 | 543 | ||||
XM_011513876.2 | XP_011512178.1 | 2 | 427 | |||
XM_017008543.1 | XP_016864032.1 | 2 | 427 | |||
XM_017008544.1 | XP_016864033.1 | 2 | 377 | |||
GenBank mRNA | AB013299.1 | - | 2 | 417 | ||
AF235023.1 | - | 2 | 346 | |||
AF331796.1 | - | 2 | 454 | |||
AK022512.1 | - | 2 | 374 | |||
AK023147.1 | - | 1 | 124 | |||
BC101476.1 | - | 2 | 331 | 8068982 | ||
BX474610.1 | - | 2 | 370 |