Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127895.1 | NP_001121367.1 | 3, 4 | 643 | ||
NM_001127896.1 | NP_001121368.1 | 2, 3 | 554 | |||
NM_022467.3 | NP_071912.2 | 2, 3 | 896 | |||
XM_011527222.1 | XP_011525524.1 | 2, 3 | 877 | |||
XM_011527224.1 | XP_011525526.1 | 3, 4 | 1011 | |||
XM_017027143.1 | XP_016882632.1 | 2, 3 | 547 | |||
GenBank mRNA | AB047801.1 | - | 2, 3 | 410 | ||
AF300612.1 | - | 2, 3 | 357 | |||
AF305781.1 | - | 2, 3 | 453 | |||
BC011380.1 | - | 2, 3 | 506 | 3874578 | ||
BC014250.2 | - | 2, 3 | 550 | 4635405 | ||
BC018723.1 | - | 2, 3 | 896 | 4137487 | ||
BI917117.1 | - | 2, 3 | 554 | |||
BX354555.2 | - | 3, 4 | 642 |